Lithium standard solution, for ICP, in HNO₃, CRM, VHG
Toimittaja: LGC Standards PROMOCHEM
VHG-LLIN-100 VHG-PLIN-100 VHG-PLIN-500 VHG-TLIN-100 VHG-TLIN-500
PROOVHG-LLIN-100EA 90.8 EUR
PROOVHG-LLIN-100 PROOVHG-PLIN-100 PROOVHG-PLIN-500 PROOVHG-TLIN-100 PROOVHG-TLIN-500
Lithium standard solution, for ICP, in HNO₃, CRM, VHG
Standardiaineet ja vertailumateriaalit Cationic Standards Lithium Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifikaatit: Traceable to NIST, SRM.