Indium standard solution, for ICP, in 5% HNO₃, VHG
Toimittaja: LGC Standards PROMOCHEM
VHG-PINN-100 VHG-PINN-500 VHG-TINN-100 VHG-TINN-500
PROOVHG-PINN-100EA 95.7 EUR
PROOVHG-PINN-100 PROOVHG-PINN-500 PROOVHG-TINN-100 PROOVHG-TINN-500
Indium standard solution, for ICP, in 5% HNO₃, VHG
Standardiaineet ja vertailumateriaalit Cationic Standards Indium Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifikaatit: Traceable to NIST, SRM.