Mercury standard solution, for ICP, in HNO₃, CRM, VHG
Toimittaja: LGC Standards PROMOCHEM
VHG-PHGN-100 VHG-PHGN-500 VHG-THGN-100 VHG-THGN-500 VHG-LHGN-100 VHG-LHGN-250
PROOVHG-PHGN-100EA 82.5 EUR
PROOVHG-PHGN-100 PROOVHG-PHGN-500 PROOVHG-THGN-100 PROOVHG-THGN-500 PROOVHG-LHGN-100 PROOVHG-LHGN-250
Mercury standard solution, for ICP, in HNO₃, CRM, VHG
Standardiaineet ja vertailumateriaalit Cationic Standards Mercury Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifikaatit: Traceable to NIST, SRM.