Aluminium standard solution, for ICP, in 5% HNO₃, VHG
Toimittaja: LGC Standards PROMOCHEM
VHG-PALN-100 VHG-PALN-500 VHG-TALN-100 VHG-TALN-500 VHG-LALN-100
PROOVHG-PALN-100EA 82.5 EUR
PROOVHG-PALN-100 PROOVHG-PALN-500 PROOVHG-TALN-100 PROOVHG-TALN-500 PROOVHG-LALN-100
Aluminium standard solution, for ICP, in 5% HNO₃, VHG
Standardiaineet ja vertailumateriaalit Cationic Standards Aluminium Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifikaatit: Traceable to NIST, SRM.