Tellurium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Toimittaja: LGC Standards PROMOCHEM
VHG-PTEN-100 VHG-PTEN-500
PROOVHG-PTEN-100EA 128 EUR
PROOVHG-PTEN-100 PROOVHG-PTEN-500
Tellurium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standardiaineet ja vertailumateriaalit Cationic Standards Tellurium Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifikaatit: Traceable to NIST, SRM.