Erbium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Toimittaja: LGC Standards PROMOCHEM
VHG-PERN-100 VHG-PERN-50 VHG-PERN-500 VHG-TERN-100 VHG-TERN-500
PROOVHG-PERN-100EA 97.4 EUR
PROOVHG-PERN-100 PROOVHG-PERN-50 PROOVHG-PERN-500 PROOVHG-TERN-100 PROOVHG-TERN-500
Erbium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standardiaineet ja vertailumateriaalit Cationic Standards Erbium Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifikaatit: Traceable to NIST, SRM.