Beryllium standard solution, for ICP, in HNO₃, CRM, VHG
Toimittaja: LGC Standards PROMOCHEM
VHG-PBEN-100 VHG-PBEN-500 VHG-TBEN-100 VHG-TBEN-500 VHG-LBEN-100
PROOVHG-PBEN-100EA 94.1 EUR
PROOVHG-PBEN-100 PROOVHG-PBEN-500 PROOVHG-TBEN-100 PROOVHG-TBEN-500 PROOVHG-LBEN-100
Beryllium standard solution, for ICP, in HNO₃, CRM, VHG
Standardiaineet ja vertailumateriaalit Cationic Standards Beryllium Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifikaatit: Traceable to NIST, SRM.