Lead standard solution, for ICP, in HNO₃, VHG
Toimittaja: LGC Standards PROMOCHEM
VHG-PPBN-100 VHG-PPBN-500 VHG-LPBN-100 VHG-LPBN-500 VHG-PPBN-50 VHG-TPBN-100 VHG-TPBN-500
PROOVHG-PPBN-100EA 82.5 EUR
PROOVHG-PPBN-100 PROOVHG-PPBN-500 PROOVHG-LPBN-100 PROOVHG-LPBN-500 PROOVHG-PPBN-50 PROOVHG-TPBN-100 PROOVHG-TPBN-500
Lead standard solution, for ICP, in HNO₃, VHG
Standardiaineet ja vertailumateriaalit Cationic Standards Lead Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Sertifikaatit: Traceable to NIST, SRM.