Nickel standard solutions
Toimittaja: LGC Standards PROMOCHEM
NIST-3136
PROONIST-3136EA 1770 EUR
PROONIST-3136
Nickel standard solutions
Standardiaineet ja vertailumateriaalit Cationic Standards Nickel Standards
This reference standard material is intended for use as a primary calibration standard for the quantitative determination of nickel.
A unit consists of five 10 ml sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of nickel.
Sertifikaatit: Traceable to NIST and SRM numbers.