Cadmium(II) cyclohexanebutyrate

Toimittaja: LGC Standards PROMOCHEM
Varoitus

NIST-1053A
PROONIST-1053AEA 1340 EUR
PROONIST-1053A
Cadmium(II) cyclohexanebutyrate
Cadmium(II) cyclohexanebutyrate
Kaava: C₂₀H₃₄CdO₄
MW: 450.9 g/mol
MDL numero: MFCD00036398
CAS-numero: 55700-14-6
EINECS: 259-767-7
UN: 2570
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR