Amitratsi

Toimittaja: LGC Standards PROMOCHEM

BP015
PROOBP015EA 492 EUR
PROOBP015
Amitratsi
Amitratsi
Kaava: C₁₉H₂₃N₃
MDL numero: MFCD00069396
CAS-numero: 33089-61-1
EINECS: 251-375-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR