Slide Printer, Signature
Toimittaja: Primera Technology Europe
078108
PRMA078108EA 12110 EUR
PRMA078108
Slide Printer, Signature
Tulostimet ja tulostimien lisävarusteet Cassette and Slide Printers
The Signature slide printer can significantly increase laborator efficiency while helping to reduce the risk of incorrect sample identification. It prints directly onto slides, eliminating handwriting errors and the need for difficult to apply, xylene-resistant labels.
  • Efficient, hands-free operation
  • Slides are stored in easy-to-load cartridges away from dust and other potential contaminants
  • USB interface enables integration with laboratory information systems
  • Chemical resistant ink ensures reliable identification of slides
With 300 dpi printMore Product Information