Caro XT-R DOFA Cleaning Trolley
Toimittaja: PFENNIG REINIGUNGSTECHNIK
7992000
PPSA7992000EA 236 EUR
PPSA7992000
Caro XT-R DOFA Cleaning Trolley
Kuljetuskärryt, siivousvaunut
Compact double trolley attractive appearance made of recycled material.
  • Cleaning method: vertical press
  • Plastic components from recycled material
  • Minimum space requirement with maximum functionality
  • Workspace: One vertical press XTR and two bucket XTR 25 L
Basic components: Basic module symmetrical (plastic made from recycled material) and swivel castors 80 mm. The trolley should be completely cleaned after use and stored in a dry place. Store EasyMop system boxes open.
Pakkaus: Packing unit: 1; carton size: 560×470×740 mm; Carton weight: 10.69 kg.