Hand brushes and brooms, PP
Toimittaja: PFENNIG REINIGUNGSTECHNIK
3590300
PPSA3590300EA 36.3 EUR
PPSA3590300
Hand brushes and brooms, PP
Harjat siivoukseen
White hand broom with run direction for hygienically sensitive areas to clean difficult-to-access areas in cleanrooms. Material: PP (body), PBT (trimming).
  • Stock height 45 mm
  • Unslit fiber ends
  • Hard bristle material