Cleanroom Sword, CR/A
Toimittaja: PFENNIG REINIGUNGSTECHNIK
3500951
PPSA3500951EA 72.6 EUR
PPSA3500951
Cleanroom Sword, CR/A
Siivousvälineet
Flexible holder for cleaning hard-to-reach places and narrow gaps.
  • Autoclavable
  • ClinoConnect Lock handle
  • Locking mechanism
  • Cleaning of hard-to-reach areas and narrow gaps
Grip material: Polyamide, glass-fibre reinforced
Fin material: Plastic Purell, FE silicon manganese wire