Cleanroom mops, MicroSicuro
Toimittaja: PFENNIG REINIGUNGSTECHNIK
2700535 2700246
PPSA2700535EA 22.8 EUR
PPSA2700535 PPSA2700246
Cleanroom mops, MicroSicuro
Siivousvälineet
Mops for all smooth and textured hard floorings and safety tiles. Made up of 100% PES super microfiber with soft-brush.
  • Ready-to-use pre-washed and excellent cleaning performance
  • With backfilling for enormous sqm-performance and cover sheet
  • Tear proof pocket of harness
Germ reduction of bacteria, yeast fungus and mould of over 99 per cent will be reached by using sterile water of standardized hardness (Testing method DGMH 2001. standard method 14).
For the use in ISO class 9.