Cleaning trolley Clino®, STEC7, with multiBox and plusBox
Toimittaja: PFENNIG REINIGUNGSTECHNIK
2088622 2088620 2088640 3077670 2088611 2088644 2088666 2088677 2088633 2088660 2088610
PPSA2088622EA 20.4 EUR
PPSA2088622 PPSA2088620 PPSA2088640 PPSA3077670 PPSA2088611 PPSA2088644 PPSA2088666 PPSA2088677 PPSA2088633 PPSA2088660 PPSA2088610
Cleaning trolley Clino®, STEC7, with multiBox and plusBox
Kuljetuskärryt, siivousvaunut
The STEC ergonomically optimised system trolley is exclusively developed and used for controlled areas. It is made of stainless steel, has enclosed appearance and open function with Clino® MultiBox and PlusBox.
  • Alternating between one and two-stage process is possible
  • Floor cleaning is simple with flat wringer Ringo and mop-reuse-technique
  • MultiBox with enormous storage space, extendable and lockable
  • PluBox system for the easy impregnation of mop covers and cloth (suitable for 6 l boxes)
  • Disposal unit at rest can be folded in, every time exchangeable and