Pierce 3UV UV Lamp
Toimittaja: Thermo Fisher Scientific
95035
PIER95035EA 948 EUR
PIER95035
Pierce 3UV UV Lamp
Valot UV-lamput
This compact ultraviolet lamp is intended for longwave, midrange and shortwave UV applications (252 nm, 302 nm and 365 nm). The lamp's innovative design allows users to dial-in the required wavelength with just the turn of a knob. The lamp is compatible with photo-reactive crosslinking and label-transfer reagents, including the Photo-reactive Amino Acids for discovering in-cell protein interactions.
  • Three wavelengths, 254/302/365 nm UV, in one lamp
  • Selection of each wavelength is by an easy turn of the dial located on one end of the housing
  • The uniquely