Isobaric mass tagging kits and reagent sets, TMT™
Toimittaja: Thermo Fisher Scientific
90063 90064 90065 90066 90068 90060 90061 90062
PIER90063EA 923 EUR
PIER90063 PIER90064 PIER90065 PIER90066 PIER90068 PIER90060 PIER90061 PIER90062
Isobaric mass tagging kits and reagent sets, TMT™
Proteiinien puhdistusjärjestelmät Protein Extraction Kits
Pierce™ TMT™ Isobaric Mass Tagging kits and reagents enable multiplex quantitation of proteins extracted from cells and tissues using tandem mass spectrometry. The Tandem Mass Tag (TMT) reagent family consists of TMTduplex and TMTsixplex sets which are specially designed to enable a rapid and cost-effective transition from method development to high-throughput protein quantitation.
  • Ideal for quantitative analysis of proteins for which no antibodies are available
  • Used in identification and quantitation of membrane and post-translationally modified proteins
Each mass-tagging reagent within a set has the same nominal mass (ie. isobaric) and chemical structure composed of an amine-reactive NHS-ester group, a spacer arm and an MS/MS reporter. The reagent sets can be used to label two or six peptide samples prepared from cells or tissues. For each sample, a unique reporter in the low mass region of the MS/MS spectrum (ie. 126-127 Da for TMTduplex and 126-131 Da for TMTsixplex Isobaric Label Reagents) is used to measure relative protein expression levels during peptide fragmentation.
Toimitustiedot: Kits are supplied as standalone sets or in optimised kit formats containing all necessary reagents and controls for maximum flexibility.