Protein magnetic beads, Protein A, Protein G, Pierce™
Toimittaja: Thermo Fisher Scientific
88847 88846 88845 88848
PIER88847EA 256 EUR
PIER88847 PIER88846 PIER88845 PIER88848
Protein magnetic beads, Protein A, Protein G, Pierce™
Helmet ja partikkelit Magnetic Beads
These protein magnetic beads are high-capacity and high-throughput affinity particles for antibody purification and immunoprecipitation methods using manual or robotic magnetic separators.
  • Higher efficiency in yield of IP target antigens
  • Low non-specific binding of stable, pre-blocked beads provide highly purified product
  • Magnetic beads eliminate resin loss and provide for more efficient separation than traditional IP methods that use only centrifugation
  • Beads are compatible with manual and automated workflows making it versatile
Recombinant protein,More Product Information