Spin filter plates, 96 well, for Ni-NTA proteins, HisPur™
Toimittaja: Thermo Fisher Scientific
88230
PIER88230EA 524 EUR
PIER88230
Spin filter plates, 96 well, for Ni-NTA proteins, HisPur™
Kuoppalevyt Suodatinlevyt
The HisPur™ Ni-NTA spin plates contain a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.
  • High capacity binding up to 60 mg of 6xHis-tagged protein per ml of resin
  • Purify proteins using native or denaturing conditions
  • Resin can be reused at least five times without reduction in performance
  • Purifies high levels of overexpressed His-tagged fusion proteins from bacterial lysates
These spin plates are ideal for simultaneous processing of multiple small (1 mg/well binding capacity) samples.More Product Information