Western blotting detection reagents, ECL western blotting substrate
Toimittaja: Thermo Fisher Scientific
32109 32209 32106
PIER32109EA 72.1 EUR
PIER32109 PIER32209 PIER32106
Western blotting detection reagents, ECL western blotting substrate
Elektroforeesireagenssit Western Blotting Reagents
ECL™ Western blotting substrate is an entry-level Western blotting substrate.
  • Less cost than other ECL substrates
  • Switch from other entry-level ECL substrates without the need for optimisation or protocol changes