Pierce™ Disposable Columns
Toimittaja: Thermo Fisher Scientific
29920 29922 29925 29924
PIER29920EA 368 EUR
PIER29920 PIER29922 PIER29925 PIER29924
Pierce™ Disposable Columns
Kromatografiakolonnit
Empty polystyrene columns with matching frits and caps for use in a variety of resin based affinity or desalting techniques.
  • For benchtop gravity flow affinity purification and size exclusion chromatography
  • Supplied complete with porous polyethylene discs, stoppers and end caps
  • Compatible with most types of aqueous buffer eluents
  • Plastic funnels can be attached to expand the buffer reservoir for increased flow rates