Natriumlauryylisulfaatti, Pierce™

Toimittaja: Thermo Fisher Scientific
Vaara

28365 28364
PIER28365EA 339 EUR
PIER28365 PIER28364
Natriumlauryylisulfaatti, Pierce™
Natriumlauryylisulfaatti

Thermo Scientific Sodium Dodecyl Sulfate (Lauryl) is standard-grade SDS detergent for use in protein polyacrylamide gel electrophoresis (PAGE).


  • Popular anionic detergent for a variety of protein methods
  • Especially useful for denaturing polyacrylamide gel electrophoresis
  • Common component of cell lysis buffers


Properties of SDS (values for pure C12):
Detergent Class: Ionic (anionic)
Aggregation Number: 62
Micelle Molecular Weight: 18,000 g
Critical Micelle Concentration (CMC): 6 to 8 mM (0.1728 to 0.2304%, w/v)
Cloud Point: >100°C
Dialyzable: No


Tilaustiedot: Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.


Varoitus: For Research Use Only. Not for use in diagnostic procedures.

Kaava: CH₃(CH₂)₁₁OSO₃Na
Sulamispiste: 204…207 °C
Tiheys: 1,1 g/cm³ (20 °C)
Leimahduspiste: >100 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036175
CAS-numero: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR