Pierce™ Protein A Agarose
Toimittaja: Thermo Fisher Scientific
20333 20334 20356
PIER20333EA 577 EUR
PIER20333 PIER20334 PIER20356
Pierce™ Protein A Agarose
Proteiininpuhdistusjärjestelmät Protein Purification Resins
Pierce™ Protein A agarose is a standard-capacity Protein A beaded agarose resin for use in a variety of antibody affinity purification methods.
  • Immobilised Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein A is immobilised by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • Standard capacity beads with a normal load of immobilised Protein A
Binding capacity: 12 to 19 mg human IgG/ml resin (approx. 6 to 8 mg mouse IgG/ml resin)