Agarose I, Pierce™
Toimittaja: Thermo Fisher Scientific
17850 17852
PIER17850EA 364 EUR
PIER17850 PIER17852
Agarose I, Pierce™
Elektroforeesigeelit Agaroosi ja akryyliamidi
Agarose I (molecular biology grade) is a highly purified and carefully blended formulation that provides uniform lot-to-lot consistency for preparation of electrophoresis gels.
  • Standard melting temperature agarose for electrophoresis
  • Separation range: 100 bp to >30 kb