Pierce™ Immunoassay Microplates, Streptavidin Coated, 384-Well
Toimittaja: Thermo Fisher Scientific
15405 15407 15504 15505 15506
PIER15405EA 581 EUR
PIER15405 PIER15407 PIER15504 PIER15505 PIER15506
Pierce™ Immunoassay Microplates, Streptavidin Coated, 384-Well
Kuoppalevyt Mikrolevyt immunologiaan
Pierce™ streptavidin coated PS microplates are ideal for binding assays using biotinylated antibodies. High binding capacity (HBC) plates provide a wider detection range and better curve linearity for small biotinylated ligands, such as peptides and oligonucleotides, than the standard coated plates. Available in clear for colorimetric assays, white for chemiluminescent assays, and black for fluorescent assays.
  • Low non specific binding
  • Easy and gentle immobilisation of biotin-containing conjugates
  • No denaturing of the protein component of a conjugate upon