Superficially porous particle (SPP) columns, Brownlee™ C18
Toimittaja: PerkinElmer
N9308401 N9308402 N9308407 N9308408 N9308409 N9308411 N9308413 N9308415 N9308513 N9308514 N9308515
PERCN9308401EA 0 EUR
PERCN9308401 PERCN9308402 PERCN9308407 PERCN9308408 PERCN9308409 PERCN9308411 PERCN9308413 PERCN9308415 PERCN9308513 PERCN9308514 PERCN9308515
Superficially porous particle (SPP) columns, Brownlee™ C18
Kromatografiakolonnit
These superficially porous particle (SPP) columns produce sharper peaks and faster separation results due to their breakthrough particle design and size.
  • General purpose octadecyl phase for reversed phase separations
  • Exhibits peak shape for a wide range of compounds
  • pH range of 2 to 9 and the temperature range of 60 °C
The design allows for a shorter diffusion path, reducing the time solute molecules spend inside the particles while passing through the stationary phase.