Thermal desorber tubes
Toimittaja: PerkinElmer
N9307050 N9307051 N9307052 N9307053 N9307054 N9307055 N9307056 N9307058 N9307061 N9307063 N9307064 N9307065
PERCN9307050EA 2070 EUR
PERCN9307050 PERCN9307051 PERCN9307052 PERCN9307053 PERCN9307054 PERCN9307055 PERCN9307056 PERCN9307058 PERCN9307061 PERCN9307063 PERCN9307064 PERCN9307065
Thermal desorber tubes
Kromatografiaan tarvikkeet Kromatografiaan termodesorptioputket
These desorber tubes are offered with a variety of sorbent packing materials for many GC applications including indoor and outdoor air monitoring, analysis of flavors and fragrances and the analysis of outgassing from packaging, polymers, pharmaceuticals and semi conductor material.
  • Each tube maintains its unique serial number which is etched for easy identification
  • With plastic end caps for short-term storage