SPE cartridges and columns, Supra-Clean®, C18
Toimittaja: PerkinElmer
N9306448 N9306422 N9306430 N9306438 N9306462 N9306475 N9306476 N9306478 N9306479 N9306523 N9306587 N9306588 N9306589
PERCN9306448EA 169 EUR
PERCN9306448 PERCN9306422 PERCN9306430 PERCN9306438 PERCN9306462 PERCN9306475 PERCN9306476 PERCN9306478 PERCN9306479 PERCN9306523 PERCN9306587 PERCN9306588 PERCN9306589
SPE cartridges and columns, Supra-Clean®, C18
Kromatografiakolonnit
  • Pure spherical silica
  • Pore size 60 to 120 Å
  • pH range 2 to 8