Calibration standard, single element, Lithium for AAS and ICP-OES, pure grade
Toimittaja: PerkinElmer
N9303781
PERCN9303781EA 95.1 EUR
PERCN9303781
Calibration standard, single element, Lithium for AAS and ICP-OES, pure grade
Standardiaineet ja vertailumateriaalit Cationic Standards Lithium Standards
Pure Grade Standards for AA and ICP-OES
  • Analyzed by ICP-OES
  • Analyzed by Classical Wet Assay
  • 32 trace impurities analyzed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppm level
  • All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Varoitus: These standards are in acid solution, for full details of hazards and specification, please see website and the links to Material Safety Data Sheets (MSDS) and Technical Data sheets.