Macro cuvettes for spectrophotometers
Toimittaja: PerkinElmer
B0631009 B0631002 B0631003 B0631004 B0631005 B0631006 B0631008 B0631010 B0631011 B0631012 B0631014 B0631015 B0631017 B0631021 B0631025 B0631026 B0631027 B0631028 B0631029 B0631030 B0631031 B0631032 B0631033 B0631034 B0631035 B0631036
PERCB0631009EA 457 EUR
PERCB0631009 PERCB0631002 PERCB0631003 PERCB0631004 PERCB0631005 PERCB0631006 PERCB0631008 PERCB0631010 PERCB0631011 PERCB0631012 PERCB0631014 PERCB0631015 PERCB0631017 PERCB0631021 PERCB0631025 PERCB0631026 PERCB0631027 PERCB0631028 PERCB0631029 PERCB0631030 PERCB0631031 PERCB0631032 PERCB0631033 PERCB0631034 PERCB0631035 PERCB0631036
Macro cuvettes for spectrophotometers
Kyvetit Spektrofotometrin kyvetit
Standard rectangular macro cuvettes frequently used in spectroscopy for routine liquids analysis. They have cell windows that are fused to the bodies by a special process which avoids deformation of the windows. This assures that the full width of the cell is usable and the finished cell dimensions are maintained.
  • High cell transmission and accurate pathlength
  • Each cell has a standard width of 9,5 mm inside
  • Made of three different window materials
Suprasil® 300 synthetic quartz is free from OH absorption and suitable for applications in the nearMore Product Information