Depth filter capsules, Supracap™ 100
Toimittaja: Cytiva (Formerly Pall Lab)
NP6PDD11 NP5LB0206 NP5LP9001 NP5LP9006 NP5LPDD11 NP5LPDD16 NP5LPDE26 NP5LPDH41 NP5LPDH46 NP5LPDK51 NP5LPDK56 NP6B0201 NP5LB0201 NP6PDK51 NP6PDK56 NP6PDH46 NP6PDE21 NP6PDD16 NP5LP2001 NP5LP1006 NP5LP2006 NP5LP2501 NP5LP2506 NP5LP7001 NP5LP7006 NP5LP1001 NP6P2001 NP6P2006 NP6P2501 NP6P2506 NP6P7001 NP6P7006 NP6P1006 NP5LPDE21 NP5LPDP87 NP6PDP81 NP6PDP86
PALLNP6PDD11EA 462 EUR
PALLNP6PDD11 PALLNP5LB0206 PALLNP5LP9001 PALLNP5LP9006 PALLNP5LPDD11 PALLNP5LPDD16 PALLNP5LPDE26 PALLNP5LPDH41 PALLNP5LPDH46 PALLNP5LPDK51 PALLNP5LPDK56 PALLNP6B0201 PALLNP5LB0201 PALLNP6PDK51 PALLNP6PDK56 PALLNP6PDH46 PALLNP6PDE21 PALLNP6PDD16 PALLNP5LP2001 PALLNP5LP1006 PALLNP5LP2006 PALLNP5LP2501 PALLNP5LP2506 PALLNP5LP7001 PALLNP5LP7006 PALLNP5LP1001 PALLNP6P2001 PALLNP6P2006 PALLNP6P2501 PALLNP6P2506 PALLNP6P7001 PALLNP6P7006 PALLNP6P1006 PALLNP5LPDE21 514-1411 PALLNP6PDP81 PALLNP6PDP86
Depth filter capsules, Supracap™ 100
Suodattimet Membraanisuodatinkapselit
Supracap™ 100 depth filter capsules with PP housing are scalable capsules with Seitz® high performance depth filter media. They are suitable for process development and pilot scale applications.
  • Seamless linear scalability
  • Completely disposable
  • Encapsulated design reduces operator exposure to potential biohazards
Low hold-up volume allows for increased product recovery and requires low post-use rinse volumes.
Applications include biopharmaceuticals, mammalian cell cultures, therapeutic protein, vaccines, blood plasma proteins and serum, yeast, E.More Product Information