Desiccator cabinets
Toimittaja: Palbam
UCD805050 UCD-604040 UCD-604060
PALBUCD805050EA 0 EUR
PALBUCD805050 PALBUCD-604040 PALBUCD-604060
Desiccator cabinets
Eksikaattorit ja kaappieksikaattorit Kaappieksikaattori
Bench top N₂ purge desiccator cabinets made from stainless steel 304, units have an electropolished finish and static dissipative PVC window. Cabinets cut nitrogen expenses by up to 80%, automate clean, dry benchtop storage eliminating moisture & oxygen related degradation and optimise yields. An appropriate flow of nitrogen is introduced into the upper section of the desiccator; the heavier air is then purged out of the bottom. Ideal for use with semiconductor components, biological and pharmaceutical samples, and other sensitive materials. Fully integrated turnkey system takes 30 s toMore Product Information