Fluoresoivat nukleotidit, Amersham CyDye™
Toimittaja: Cytiva
PA53521 PA53021 PA53022 PA55021 PA55022 PA53026 PA55026 PA55321 PA55322 PA53031
PA53521EA 521 EUR
PA53521 PA53021 PA53022 PA55021 PA55022 PA53026 PA55026 PA55321 PA55322 PA53031
Fluoresoivat nukleotidit, Amersham CyDye™
Proteiinien yhdistämis- ja leimausjärjestelmät
Fluorescent nucleotides for labelling DNA and RNA probes, especially during multi-colour analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.
  • High sensitivity and low non specific binding
  • High photostability, high water solubility
  • pH insensitive
CyDye™ labelled nucleotides are available in a range of bright and intense colours with narrow emission bands for multiplexing with a single sample.