Phenol red, sodium salt
Toimittaja: BIOWEST
P5648-10GR
P5648-10GREA 51.5 EUR
P5648-10GR
Phenol red, sodium salt
Soluviljelyyn suplementit ja lisäaineet
Phenol red is used as a pH indicator in cell culture. Its colour exhibits a gradual transition from yellow to red over the pH range 6,8 to 8,2. Above pH 8,2, phenol red turns a bright pink (fuchsia) colour.