1,3,5-Trihydroksibentseeni, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY
Varoitus

Synonyms: 1,3,5-Trihydroxybenzene

M-Clarity™ quality level = MQ100

P3502-100G P3502-25G
P3502-100GEA 176 EUR
P3502-100G P3502-25G
1,3,5-Trihydroksibentseeni, Sigma-Aldrich®
1,3,5-Trihydroksibentseeni

Phloroglucinol (phlo) is a phenol derivative that shows a cytoprotective effect from oxidative damage by enhancing the activity of cellular catalase. It can also be used to prepare synthetic analogs of A-type proanthocyanidins (PACs) such as 2,8-dioxabicyclo nonane derivatives by reacting with the corresponding flavylium salts. Phlo can react with benzaldehyde derivatives to form phloroglucinol-based microporous polymeric organic frameworks (phlo-POF) with potential applications in ion exchange and gas adsorption. Phloroglucinol is an antispasmodic and a derivative of phenol.


  • Phloroglucinol has been used to prevent hyperhydricity by promoting the lignification of shoots
  • It has also been used in PG (phloroglucinol)-sucrose treatments for the growth and development of in vitro derived shoot tips
  • Phloroglucinol has been used to stain root sections. It has also been used to stain plant sections to identify plant materials like lignin


Phloroglucinol displays antispasmodic activity. It is a trihydroxybenzene with antithrombotic, profibrinolytic, antimicrobial, antimalarial, cancer chemopreventive, anti-HIV, and anti-leishmanial activities. Phloroglucinol (PG) is a biosynthetic precursor of the 2,4-diacetylphloroglucinol (DAPG) an antibiotic against soil-borne diseases. Phloroglucinol is a useful intermediate because it is polyfunctional.

Kaava: C₆H₆O₃
Kiehumispiste: 400 °C (1013 hPa)
Sulamispiste: 215…220 °C
Tiheys: 1,46 g/cm³ (20 °C)
Storage Temperature: Room Temperature
MDL numero: MFCD00002286
CAS-numero: 108-73-6
EINECS: 203-611-2
Merck Index: 12,07482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=856fe9381dd782126a6e308dbad39b1b03a9c177&vid=4ff8d9858e288b879cbb1fc143d2bb1821e802c9&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance (colour) White to faint yellow to faint beige
Appearance (form) Powder or crystals
Purity (HPLC area %) Report result
Solubility (colour) Colorless to light brown-yellow
Solubility (turbidity) Clear to hazy
Solubility (method) 50 mg/ml ethanol
Loss on drying Report result
Carbon content Report result
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR