DNA transfection reagent, CombiMag™
Toimittaja: OZ BIOSCIENCES
CM20100 CM20200 CM21000 KC30200
OZBICM20100EA 70.95 EUR
OZBICM20100 OZBICM20200 OZBICM21000 OZBIKC30200
DNA transfection reagent, CombiMag™
Transfektioreagenssit
CombiMag™ transfection reagent is based on the use of magnetic nanoparticles for improving transfection reagent efficiency. It has been designed to be employed in association with any commercial transfection reagent and can be used with all types of nucleic acids. It has been successfully tested on a broad range of primary cells, hard to transfect cells and cell lines.
  • Ideal to enhance any transfection reagent efficiency without changing the protocol/cells
  • Perfect for primary cells and hard-to-transfect cells
  • Increase any transfection reagent efficiency
  • Non toxic
  • Compatible with and without serum
  • Simple, rapid and easy to use
Store at +4 °C.
Lisävarusteiden tiedot: This reagent needs to be used with a magnetic plate.
Toimitustiedot: Shipped at room temperature.