Oxoid biochemical identification system (O.B.I.S.)
Toimittaja: THERMO FISHER DIAGNOSTICS
ID0570M ID0580M ID0600M
OXOIID0570MEA 297 EUR
OXOIID0570M OXOIID0580M OXOIID0600M
Oxoid biochemical identification system (O.B.I.S.)
Pikatestit patogeenien ja allergeenien osoittamiseen Enzymatic Tests
The Oxoid Biochemical Identification System (O.B.I.S.) is a rapid colorimetric test for the determination of biochemical activity in microorganisms. It is a rapid screening method for differentiation of other species.
  • Reduces the need for full biochemical identification of all suspect colonies