Latex tests, Dryspot
Toimittaja: THERMO FISHER DIAGNOSTICS
DR0100M DR0500G DR0585A DR0586G DR0587G DR0588G DR0589G DR0590G DR0591G DR0592G DR0593G DR0595A DR0800M DR0801M DR0802M DR0803M DR0850M DR0900A DR0420M DR0850B DR0220M DR1108A DR0200M DR0210M
OXOIDR0100MEA 239 EUR
OXOIDR0100M OXOIDR0500G OXOIDR0585A OXOIDR0586G OXOIDR0587G OXOIDR0588G OXOIDR0589G OXOIDR0590G OXOIDR0591G OXOIDR0592G OXOIDR0593G OXOIDR0595A OXOIDR0800M OXOIDR0801M OXOIDR0802M OXOIDR0803M OXOIDR0850M OXOIDR0900A OXOIDR0420M OXOIDR0850B OXOIDR0220M OXOIDR1108A OXOIDR0200M OXOIDR0210M
Latex tests, Dryspot
Pikatestit patogeenien ja allergeenien osoittamiseen Immunologiset pikatestit Ag-AK, Latex
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
  • Simplified room temperature storage allows storage for up to two years
  • Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards