Filling solutions for pH electrodes, Orion™
Toimittaja: Thermo Orion
900003 900019 510011 810007 900001 900002 900004 900011 910008-WA
662-1524EA 227 EUR
662-1524 ORIO900019 662-1555 662-0717 662-0749 662-0750 662-0751 662-0753 ORIO910008-WA
Filling solutions for pH electrodes, Orion™
Elektrolyytit Elektrodin täyttöliuokset
Filling solutions for pH and reference electrodes.
  • ROSS filling solution for use only with ROSS™ and ROSS Ultra™ pH electrodes to prevent damaging the ROSS electrodes
  • When the single junction reference electrode or double junction reference electrodes are used with Orion ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide