Anti-VSNL1 Mouse Monoclonal Antibody [clone: UMAB115]

Toimittaja: OriGene
UM800034
ORIGUM800034EA 847 EUR
ORIGUM800034
Anti-VSNL1 Mouse Monoclonal Antibody [clone: UMAB115]
Vasta-aineet
Recommended Dilutions: Immunochemistry: 1:200

Type: Primary
Antigen: VSNL1
Clonality: Monoclonal
Clone: UMAB115
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR