Human ERBB2
Toimittaja: OriGene
TS422909P5
ORIGTS422909P5EA 1630 EUR
ORIGTS422909P5
Human ERBB2
Proteiinit
HEK293T cells were transiently transfected with TrueORF cDNA plasmid. Transfected cells were cultured for 48 hrs. After harvesting, the cultured cells were fixed in formalin and dehydrated before embedding in paraffin. 5 µm sections of the FFPE cell pellet blocks are cut and mounted on positively charged SuperFrost slides.
  • Expression host: HEK293T
Stability: Slides are guaranteed for a year from the date of receipt if proper storage instructions were followed
Varoitus: This product is for research use only and is not approved for use in humans or in clinical diagnosis.