Anti-BOLL Mouse Monoclonal Antibody [clone: 4C2]

Toimittaja: OriGene
TA804111
ORIGTA804111EA 621 EUR
ORIGTA804111
Anti-BOLL Mouse Monoclonal Antibody [clone: 4C2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: BOLL
Clonality: Monoclonal
Clone: 4C2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR