Anti-DOT1L Mouse Monoclonal Antibody [clone: 2B10]

Toimittaja: OriGene
TA802479
ORIGTA802479EA 621 EUR
ORIGTA802479
Anti-DOT1L Mouse Monoclonal Antibody [clone: 2B10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: DOT1L
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR