Anti-WWC1 Mouse Monoclonal Antibody [clone: OTI1B1]

Toimittaja: OriGene
TA801574
ORIGTA801574EA 621 EUR
ORIGTA801574
Anti-WWC1 Mouse Monoclonal Antibody [clone: OTI1B1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: WWC1 (WW and C2 domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR