Anti-SERPINB1 Mouse Monoclonal Antibody [clone: OTI2B10]

Toimittaja: OriGene
TA800117
ORIGTA800117EA 621 EUR
ORIGTA800117
Anti-SERPINB1 Mouse Monoclonal Antibody [clone: OTI2B10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: SERPINB1 (serpin peptidase inhibitor, clade B (ovalbumin), member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR