Anti-MS4A1 Mouse Monoclonal Antibody [clone: 3C4]

Toimittaja: OriGene
TA600506
ORIGTA600506EA 636 EUR
ORIGTA600506
Anti-MS4A1 Mouse Monoclonal Antibody [clone: 3C4]
Vasta-aineet
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR