Anti-CD5 Mouse Monoclonal Antibody [clone: 5D4]

Toimittaja: OriGene
TA600466
ORIGTA600466EA 636 EUR
ORIGTA600466
Anti-CD5 Mouse Monoclonal Antibody [clone: 5D4]
Vasta-aineet
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 5D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR