Anti-PARVA Mouse Monoclonal Antibody [clone: OTI3H9]

Toimittaja: OriGene
TA506003
ORIGTA506003EA 621 EUR
ORIGTA506003
Anti-PARVA Mouse Monoclonal Antibody [clone: OTI3H9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PARVA (parvin, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR